missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folinic acid calcium salt hydrate, ≥90%, MilliporeSigma™ Supelco™
Folinic acid calcium salt hydrate is a drug, widely used for the treatment of cancer, since it can reduce toxic side effects and helps to improve drug accumulation in tumor tissues.
Supplier: Merck Emd Millipore F7878500MG
Description
Featured Industries: Pharmaceutical (small molecule)Specifications
| 1492-18-8 (anhydrous) | |
| C20H21 CaN7O7 · xH2 O | |
| 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt; 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt; 5-HCO-H4 PteGlu; Calcium folinate; Citrovorum factor calcium salt; Leucovorin calcium salt | |
| 500 mg | |
| ≥90% |
| Neat | |
| 240°C to 250°C | |
| Analytical Standard | |
| 511.50 (anhydrous basis) | |
| Analytical Standard |