missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Folinic acid, calcium salt, 99.4%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219150425
Specifications
Folinic acid, calcium salt | |
25mg | |
0.994 | |
calcium folinate | |
C1C(N(C2=C(N1)NC(=NC2=O)N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.[Ca+2] | |
512.516 | |
511.5 |
Yellow | |
1492-18-8 | |
C20H22CaN7O7+ | |
KVUAALJSMIVURS-ZEDZUCNESA-M | |
calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate | |
131675590 | |
Powder |
Chemical Identifiers
1492-18-8 | |
512.516 | |
calcium folinate | |
calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate |
C20H22CaN7O7+ | |
KVUAALJSMIVURS-ZEDZUCNESA-M | |
131675590 | |
C1C(N(C2=C(N1)NC(=NC2=O)N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.[Ca+2] |