missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folinic acid, calcium salt, 99.4%, MP Biomedicals™
$80.15 - $80.15
Chemical Identifiers
| CAS | 1492-18-8 |
|---|---|
| Molecular Formula | C20H21CaN7O7 |
| Molecular Weight (g/mol) | 511.51 |
| MDL Number | MFCD00006704 |
| InChI Key | KVUAALJSMIVURS-AEQORCIRNA-L |
| Synonym | calcium folinate |
| PubChem CID | 131675590 |
| SMILES | [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19150425
|
MP Biomedicals Inc
0219150425 |
25 mg |
Each for $80.15
|
|
|||||
Chemical Identifiers
| 1492-18-8 | |
| 511.51 | |
| KVUAALJSMIVURS-AEQORCIRNA-L | |
| 131675590 |
| C20H21CaN7O7 | |
| MFCD00006704 | |
| calcium folinate | |
| [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 |
Specifications
| 1492-18-8 | |
| 0.994 | |
| MFCD00006704 | |
| calcium folinate | |
| [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 | |
| 511.51 | |
| 511.5 | |
| Folinic acid, calcium salt |
| Yellow | |
| C20H21CaN7O7 | |
| 25 mg | |
| KVUAALJSMIVURS-AEQORCIRNA-L | |
| calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate | |
| 131675590 | |
| Powder |