missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin sodium salt, Thermo Scientific™

$158.47 - $521.97

Chemical Identifiers

CAS 93957-55-2
Molecular Formula C24H25FNNaO4
Molecular Weight (g/mol) 433.46
MDL Number MFCD00929076
InChI Key ZGGHKIMDNBDHJB-RPQBTBOMSA-M
Synonym fluvastatin sodium, sodium 3s,5r-7-3-4-fluorophenyl-1-propan-2-yl-1h-indol-2-yl-3,5-dihydroxyhept-6-enoate
PubChem CID 66587062
IUPAC Name sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
SMILES [Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC458010010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC458010050
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

93957-55-2
433.46
ZGGHKIMDNBDHJB-RPQBTBOMSA-M
66587062
[Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
C24H25FNNaO4
MFCD00929076
fluvastatin sodium, sodium 3s,5r-7-3-4-fluorophenyl-1-propan-2-yl-1h-indol-2-yl-3,5-dihydroxyhept-6-enoate
sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate

Specifications

93957-55-2
C24H25FNNaO4
fluvastatin sodium, sodium 3s,5r-7-3-4-fluorophenyl-1-propan-2-yl-1h-indol-2-yl-3,5-dihydroxyhept-6-enoate
ZGGHKIMDNBDHJB-RPQBTBOMSA-M
sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
66587062
≥97.5% (HPLC)
Authentic
MFCD00929076
Solubility in water: 30g/L.
[Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
433.46
433.45
Fluvastatin sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only