missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin sodium salt

Catalog No. AC458010010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458010010 1 g
AC458010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458010010 Supplier Thermo Scientific Chemicals Supplier No. 458010010

CAS: 93957-55-2 | C24H25FNNaO4 | 433.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93957-55-2
Molecular Formula C24H25FNNaO4
Molecular Weight (g/mol) 433.46
MDL Number MFCD00929076
InChI Key ZGGHKIMDNBDHJB-RPQBTBOMSA-M
Synonym fluvastatin sodium, sodium 3s,5r-7-3-4-fluorophenyl-1-propan-2-yl-1h-indol-2-yl-3,5-dihydroxyhept-6-enoate
PubChem CID 66587062
SMILES [Na+].CC(C)N1C(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)=C(C2=CC=CC=C12)C1=CC=C(F)C=C1

Specifications

Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: 30g/L.
IUPAC Name sodium (3S,5R,6E)-7-[3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
Formula Weight 433.45
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Fluvastatin sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only