missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flurprimidol

Catalog No. AC466631000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466631000 100 mg
AC466635000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466631000 Supplier Thermo Scientific Chemicals Supplier No. 466631000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56425-91-3 | C15H15F3N2O2, C15H15F3N2O2 | 312.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56425-91-3
Molecular Formula C15H15F3N2O2

Specifications

Quantity 100 mg
Solubility Solubility aq. soln.: insoluble.
Melting Point 93.0°C to 97.0°C
Boiling Point 264.0°C
Formula Weight 312.29
InChI Key VEVZCONIUDBCDC-UHFFFAOYNA-N
SMILES CC(C)C(O)(C1=CC=C(OC(F)(F)F)C=C1)C1=CN=CN=C1
IUPAC Name 2-methyl-1-(pyrimidin-5-yl)-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
Molecular Weight (g/mol) 312.29
Chemical Name or Material Flurprimidol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid breathing dust/fume/gas/mist/vapors/spray.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only