missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flurbiprofen 98.0+%, TCI America™
$138.61 - $404.41
Chemical Identifiers
| CAS | 5104-49-4 |
|---|---|
| Molecular Formula | C15H13FO2 |
| Molecular Weight (g/mol) | 244.27 |
| MDL Number | MFCD00079303 |
| InChI Key | SYTBZMRGLBWNTM-UHFFFAOYNA-N |
| Synonym | flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum |
| PubChem CID | 3394 |
| ChEBI | CHEBI:5130 |
| IUPAC Name | 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid |
| SMILES | CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 5104-49-4 | |
| 244.27 | |
| SYTBZMRGLBWNTM-UHFFFAOYNA-N | |
| 3394 | |
| 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid |
| C15H13FO2 | |
| MFCD00079303 | |
| flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum | |
| CHEBI:5130 | |
| CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1 |
Specifications
| 5104-49-4 | |
| White | |
| MFCD00079303 | |
| 2811 | |
| SYTBZMRGLBWNTM-UHFFFAOYNA-N | |
| 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid | |
| 3394 | |
| 244.27 | |
| Crystalline Powder |
| 113°C | |
| C15H13FO2 | |
| 5 g | |
| flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum | |
| CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1 | |
| 244.27 | |
| CHEBI:5130 | |
| ≥98.0% (T,HPLC) | |
| Flurbiprofen |
Safety and Handling
RTECSNumber : DU8341000
TSCA : No