missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen 98.0+%, TCI America™

Supplier:  TCI America F037125G

 View more versions of this product

Catalog No. F037125G


Only null left

Description

Description

Specifications

Specifications

Flurbiprofen
113°C
C15H13FO2
25 g
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
244.27
CHEBI:5130
≥98.0% (T,HPLC)
5104-49-4
White
MFCD00079303
2811
SYTBZMRGLBWNTM-UHFFFAOYNA-N
2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
3394
244.27
Crystalline Powder

Chemical Identifiers

5104-49-4
244.27
SYTBZMRGLBWNTM-UHFFFAOYNA-N
3394
2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
C15H13FO2
MFCD00079303
flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
CHEBI:5130
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DU8341000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions