missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoromethyl Phenyl Sulfone 98.0+%, TCI America™
$384.00 - $1293.65
Chemical Identifiers
| CAS | 20808-12-2 |
|---|---|
| Molecular Formula | C7H7FO2S |
| Molecular Weight (g/mol) | 174.189 |
| MDL Number | MFCD00191650 |
| InChI Key | DENHPZASLKBBHA-UHFFFAOYSA-N |
| Synonym | Fluoromethylsulfonylbenzene |
| PubChem CID | 2759036 |
| IUPAC Name | fluoromethylsulfonylbenzene |
| SMILES | C1=CC=C(C=C1)S(=O)(=O)CF |
Chemical Identifiers
| 20808-12-2 | |
| 174.189 | |
| DENHPZASLKBBHA-UHFFFAOYSA-N | |
| 2759036 | |
| C1=CC=C(C=C1)S(=O)(=O)CF |
| C7H7FO2S | |
| MFCD00191650 | |
| Fluoromethylsulfonylbenzene | |
| fluoromethylsulfonylbenzene |
Specifications
| 20808-12-2 | |
| White-Yellow | |
| MFCD00191650 | |
| Fluoromethylsulfonylbenzene | |
| C1=CC=C(C=C1)S(=O)(=O)CF | |
| 174.189 | |
| 174.19 | |
| Crystalline Powder |
| 53°C | |
| C7H7FO2S | |
| 1 g | |
| DENHPZASLKBBHA-UHFFFAOYSA-N | |
| fluoromethylsulfonylbenzene | |
| 2759036 | |
| ≥98.0% (GC) | |
| Fluoromethyl Phenyl Sulfone |
Safety and Handling
TSCA : No