accessibility menu, dialog, popup

UserName

SOOF

CAS RN 20808-12-2

IUPAC Name: fluoromethanesulfonylbenzene
Synonyms: fluoromethanesulfonylbenzene
Molecular Weight (g/mol): 174.19
Molecular Formula: C7H7FO2S
InChi Key: DENHPZASLKBBHA-UHFFFAOYSA-N
SMILES: FCS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results