missing translation for 'onlineSavingsMsg'
Learn More

Fluoromethyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. F03411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03411G 1 g
F03415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03411G Supplier TCI America Supplier No. F03411G
Only null left

Chemical Identifiers

CAS 20808-12-2
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00191650
InChI Key DENHPZASLKBBHA-UHFFFAOYSA-N
Synonym Fluoromethylsulfonylbenzene
PubChem CID 2759036
IUPAC Name fluoromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CF

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fluoromethyl Phenyl Sulfone