Learn More
Fluorobenzene, 99%
CAS: 462-06-6 | C6H5F | 96.10 g/mol
Supplier: Thermo Scientific Chemicals A1438430
Description
Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
Specifications
| Fluorobenzene | |
| -42°C | |
| 84°C to 85°C | |
| Aromatic | |
| 1.466 | |
| 250 g | |
| 1236623 | |
| benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
| PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
| fluorobenzene | |
| 10008 | |
| 96.1 |
| 462-06-6 | |
| 1.025 | |
| −15°C (−5°F) | |
| C6H5F | |
| MFCD00000280 | |
| UN2387 | |
| 14,4170 | |
| Slightly soluble in water. | |
| FC1=CC=CC=C1 | |
| 96.10 | |
| CHEBI:5115 | |
| 99% |
Chemical Identifiers
| 462-06-6 | |
| 96.10 | |
| PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
| 10008 | |
| fluorobenzene |
| C6H5F | |
| MFCD00000280 | |
| benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
| CHEBI:5115 | |
| FC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P305+P351+P338-P310-P370+P378q-P501c
H225-H318
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
EINECSNumber : 207-321-7
RTECSNumber : DA0800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only