missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene, 99%

Catalog No. AAA143840B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA143840B 1000 g
AAA1438430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA143840B Supplier Thermo Scientific Chemicals Supplier No. A143840B
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 462-06-6 | C6H5F | 96.10 g/mol

Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.

Chemical Identifiers

CAS 462-06-6
Molecular Formula C6H5F
Molecular Weight (g/mol) 96.10
MDL Number MFCD00000280
InChI Key PYLWMHQQBFSUBP-UHFFFAOYSA-N
Synonym benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene
PubChem CID 10008
ChEBI CHEBI:5115
IUPAC Name fluorobenzene
SMILES FC1=CC=CC=C1

Specifications

Melting Point -42°C
Density 1.025
Boiling Point 84°C to 85°C
Flash Point −15°C (−5°F)
Odor Aromatic
Refractive Index 1.466
Quantity 1000 g
UN Number UN2387
Beilstein 1236623
Merck Index 14,4170
Solubility Information Slightly soluble in water.
Formula Weight 96.1
Percent Purity 99%
Chemical Name or Material Fluorobenzene
Show More Show Less
Hazard Category H225-H318
Hazard Statement GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
EINECSNumber 207-321-7
RTECSNumber DA0800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only