missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, Certified, 1.00ppm ±0.05ppm (1mL = 0.001mg F-), LabChem™

Catalog No. LC145602
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC145602 1 L Poly Bottle
LC145601 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC145602 Supplier LabChem Supplier No. LC145602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-49-4 , 7732-18-5
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 99.9998, 0.0002
Identification Passes Test
Physical Form Liquid
Concentration 1.00 ppm ±0.05 ppm (1mL = 0.001mg F-)
Linear Formula NaF
Color Colorless
Solubility Soluble in water
Quantity 1 L
Formula Weight 41.99
Grade Certified
Chemical Name or Material Fluoride Standard, 1ppm (1mL = 0.001mg F-)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature