missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 10 ppm F-, Ricca Chemical

Catalog No. p-3251692
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
317016 500 mL Natural Poly Bottle
317032 1 L Natural Poly Bottle
R31700004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 317016 Supplier Ricca Chemical Company Supplier No. 317016
Only null left

Chemical Identifiers

CAS 7681-49-4 , 7732-18-5
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.0022
CAS Max % 100.0, 0.0022
Equivalence 1 mL = 0.01 mg F
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Fluoride Standard