missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 0.5 ppm F- (Premixed with TISAB), Ricca Chemical

$153.91 - $153.91

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3161516
View Documents
Ricca Chemical Company
3161516
500 mL Natural Poly Bottle
Each for $153.91
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
FNa
MFCD00003524
5235
sodium fluoride

Specifications

Natural Poly Bottle
93.0,1.95,1.64,1.59,0.11
Liquid
MFCD00003524
Approximately 0°C
[F-].[Na+]
41.99
5235
1mL = 0.0005mg F-, 0.5ppm F-, Premixed with TISAB
7681-49-4,1310-73-2,64-19-7,7647-14-5,13291-61-7,7681-49-4
96.0,2.03,1.7,1.65,0.11
FNa
Colorless
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
500 mL
CHEBI:28741
Fluoride Standard
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
missing translation for 'documents'

missing translation for 'documents'