Learn More
Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
CAS: 7681-49-4 | FNa | 41.99 g/mol
Supplier: Thermo Scientific Chemicals 035550AP
Description
Fluoride, Ion chromatography standard solution, Specpure™, F∣- 1000μg/ml is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Fluoride | |
| 23 hPa (17mm Hg) at 20°C | |
| FNa | |
| Immiscible with water | |
| [F-].[Na+] | |
| 41.99 | |
| 5235 | |
| Sodium fluoride: 0.22%; Water: 99.78% |
| 7681-49-4 , 7732-18-5 | |
| Liquid | |
| MFCD00003524 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| sodium;fluoride | |
| 500 mL | |
| CHEBI:28741 |
Chemical Identifiers
| 7681-49-4 | |
| 41.99 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| CHEBI:28741 | |
| [F-].[Na+] |
| FNa | |
| MFCD00003524 | |
| 5235 | |
| sodium;fluoride |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only