missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fluorescent Shop Envelopes

Color-code work in process, machine procedures and drawings. Brady™ Fluorescent Shop Envelopes are the perfect solution for organizing work flows.

$235.16 - $251.10

Specifications

Material Plastic
Mounting Type Metal Grommet
For Use With (Application) Color-coding work in process, machine procedures and drawings
Quantity 25/Pk.
Dimensions (L x W) 12 x 9 in.
View More Specs

Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19113413
View Documents Promotion Details
Brady™
56946
Fluorescent Orange Trim on Clear
Pack of 25 Each for $235.16
Only null left
 
19113414
View Documents Promotion Details
Brady™
56948
Fluorescent Yellow Trim on Clear
Pack of 25 Each for $251.10
Only null left
 
19113412
View Documents Promotion Details
Brady™
56945
Fluorescent Green Trim on Clear
Pack of 25 Each for $251.10
Only null left
 
Description

Description

  • Fluorescent orange trimmed shop envelopes
  • Ideal for color-coding work in process, machine procedures and drawings
  • For indoor and outdoor use
Specifications

Specifications

Plastic
Color-coding work in process, machine procedures and drawings
12 x 9 in.
9 in.
Metal Grommet
25/Pk.
12 in.
Videos
SDS
Documents

Documents