missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fluorescent Shop Envelopes

Catalog No. 19113414
Change view
Click to view available options
Color:
Fluorescent Green Trim on Clear
Fluorescent Orange Trim on Clear
Fluorescent Yellow Trim on Clear
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19113414 Fluorescent Yellow Trim on Clear
19113413 Fluorescent Orange Trim on Clear
19113412 Fluorescent Green Trim on Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19113414 Supplier Brady™ Supplier No. 56948

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Color-code work in process, machine procedures and drawings. Brady™ Fluorescent Shop Envelopes are the perfect solution for organizing work flows.

  • Fluorescent yellow trimmed shop envelopes
  • Ideal for color-coding work in process, machine procedures and drawings
  • For indoor and outdoor use

Specifications

Color Fluorescent Yellow Trim on Clear
Material Plastic
Mounting Type Metal Grommet
For Use With (Application) Color-coding work in process, machine procedures and drawings
Quantity 25/Pk.
Dimensions (L x W) 12 x 9 in.
Length (English) 12 in.
Width (English) 9 in.