missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein-Poly GT
Description
A general fluorescein peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase activity.
For use in combination with the LanthaScreen™ Tb-pY20 Antibody (PV3552) to evaluate inhibitors in medium to high throughput screening applications.
For use in combination with the LanthaScreen™ Tb-pY20 Antibody (PV3552) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Amino Acid Sequence | Fluorescein- Poly Glu:Tyr (4:1) |
| Assay Entry | Biochemical terbium phospho-specific activity |
| Content And Storage | 30 μM Fluorescein Poly-GT peptide substrates in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl (HEPES buffered saline). Store at -20°C and protect from light. |
| Detection Method | Fluorescence |
| Form | Liquid |
| Quantity | 1 mL |
| Substrate | Fluorescein-Poly GT |
| Substrate Properties | Peptide-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Tyrosine Kinases |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.