missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein-MAP2K1

Catalog No. PV4812
Encompass
Change view
Click to view available options
Quantity:
20 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4812 20 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4812 Supplier Invitrogen™ Supplier No. PV4812
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescein-MAP2K1

Inactive MAP2K1 (MEK1) has been labeled with fluorescein, and is a substrate for MAPK family members.

For use in combination with the LanthaSreen™ Tb-MAP2K1 [pSer217/221] Antibody (PV4813) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage Fluorescein-MAP2K1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C.
Detection Method Fluorescence
Quantity 20 nmol
Substrate MAP2K1
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 10454, 10746, 1326, 4215, 4217, 4293, 4294, 4296, 5894, 673, 6885
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 50000 Da
Shipping Condition Dry Ice
Target Entry MAP3K2, RAF1 (cRAF) Y340D Y341D, MAP3K10, MAP3K9, MAP3K7/MAP3K7IP1, MAP3K5, MAP3K11, BRAF V599E, BRAF, MAP3K8, MAP3K3
Validated Kinases BRAF, MAP3K2, MAP3K3, MAP3K5, MAP3K7, MAP3K8, MAP3K9, MAP3K10, MAP3K11
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.