missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein-IKK Substrate
Description
A fluorescein-labeled peptide substrate that is primarily used with the IkappaB kinase (IKK) family.
For use in combination with the LanthaScreen™ Tb-IkappaBalpha [pSer32] antibody (PV3562) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Amino Acid Sequence | Fluorescein- DRHDSGLDSMKDE |
| Assay Entry | Biochemical terbium phospho-specific activity |
| Content And Storage | 1 mg/mL fluorescein-IKK substrate in water. Store at -20°C and protect from light. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Substrate | Fluorescein-IKK |
| Substrate Properties | Peptide-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Serine/Threonine Kinases |
| Gene ID (Entrez) | 1147, 2869, 3551 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.