missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein-ERM (LRRKtide)
Description
A fluorescein-labeled peptide substrate based upon ezrin/radixin/moesin (ERM) that is phosphorylated by LRRK2 and LRRK2 mutants.
For use in combination with the LanthaScreen™ Tb-anti-ERM (pLRRKtide) Antibody (PV4899) to evalute inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Amino Acid Sequence | Fluorescein-GAGRLGRDKYKTLRQIRQ |
| Assay Entry | Biochemical terbium phospho-specific activity |
| Content And Storage | 400 μM Fluorescein-ERM (LRRKtide) peptide substrate in water. Store at -20°C and protect from light. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Substrate | ERM (LRRKtide) |
| Substrate Properties | Peptide-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Serine/Threonine Kinases |
| Gene ID (Entrez) | 120892 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.