missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Diphosphate, Tetraammonium Salt (FDP)

Catalog No. F2999
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F2999 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F2999 Supplier Invitrogen™ Supplier No. F2999
  • ONLINE EXCLUSIVE PRICE
    $766.00
    $628.12 / Each

    Save $137.88

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Colorless and non-fluorescent

FDP is a colorless and nonfluorescent substrate for alkaline phosphatases. Sequential alkaline phosphatase mediated hydrolysis of its two phosphate substituents yields weakly fluorescent fluorescein monophosphate followed by strongly fluorescent fluorescein (excitation/emission approx. 490/514nm).

Alkaline Phosphatase Assays, Cell Analysis, Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 5 mg
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Alkaline Phosphatase, Tyrosine Phosphatases
Excitation/Emission 490⁄514
For Use With (Equipment) Fluorometer, Microplate Reader
Label or Dye FITC (Fluorescein)
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.