missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein-Crosstide Substrate

Catalog No. PV3509
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3509 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3509 Supplier Invitrogen™ Supplier No. PV3509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescein-Crosstide Substrate

A general substrate for Ser/Thr kinases, the Crosstide peptide substrate is based upon GSK-3alpha/beta.

For use in combination with the LanthaScreen™ Tb-pCrosstide Antibody (PV3564) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Amino Acid Sequence Fluorescein- GRPRTSSFAEG
Assay Entry Biochemical terbium phospho-specific activity
Content And Storage 1 mg/mL Fluorescein-Crosstide substrates in 50 mM HEPES, pH 7.5. Store at -20°C and protect from light.
Detection Method Fluorescence
Quantity 1 mg
Substrate Fluorescein-Crosstide
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 10000, 10298, 207, 208, 25400, 51347, 5566, 56164, 57144, 6197, 6198, 8536
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 1524 Da
Research Category Signal Transduction
Shipping Condition Dry Ice
Target Entry PAK4, TAOK3, CAMK2D, CAMK2A, PAK1, PRKACA, AKT2, RPS6KB1, AKT3, AKT1, PAK7, RPS6KA3
Validated Kinases AKT1, AKT2, AKT3, CAMK2A, CAMK2D, PAK1, PAK4, PAK7, PRKACA, RPS6KB1, RPS6KA3, TAOK3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.