missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein-CREBtide Substrate
Description
A fluorescein-labeled peptide substrate that is based upon the CREB ( cAMP responsive element binding) protein. The peptide substrate is phosphorylated by a wide range of Ser/Thr kinases.
For use in combination with the LanthaScreen™ Tb-CREB [pSer133] antibody (PV3542) to evaluate inhibitors in medium to high throughput screening applications.
For use in combination with the LanthaScreen™ Tb-CREB [pSer133] antibody (PV3542) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Amino Acid Sequence | Fluorescein- KRREILSRRPSYR |
| Assay Entry | Biochemical terbium phospho-specific activity |
| Content And Storage | 1 mg/mL Fluorescein-CREBtide substrate in water. Store at -20°C and protect from light. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Substrate | Fluorescein-CREBtide |
| Substrate Properties | Peptide-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Serine/Threonine Kinases |
| Label or Dye | FITC (Fluorescein) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.