missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Alcohol Soluble, Spectrum™ Chemical

$862.86 - $6,867.13

Chemical Identifiers

CAS 2321-07-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18610599
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $862.86
Only null left
 
18610600
View Documents Promotion Details
N/A 2.5 kg Amber Glass Bottle
Each for $6,867.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2321-07-5

Specifications

2321-07-5
Reagent
Fluorescein
1
Brown or Orange to Red
Videos
SDS
Documents

Documents

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.