missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Alcohol Soluble, Spectrum™ Chemical

Catalog No. 18610599
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610599 100 g Amber Glass Bottle
18610600 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610599 Supplier Spectrum Chemical Mfg Cor Supplier No. FL113100GM

Chemical Identifiers

CAS 2321-07-5

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Brown or Orange to Red
Quantity 100 g
Chemical Name or Material Fluorescein

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.