missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Alcohol Soluble, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Color | Brown or Orange to Red |
| Quantity | 2.5 kg |
| Molecular Weight (g/mol) | 332.31 |
| Grade | Reagent |
| Chemical Name or Material | Fluorescein |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.