missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoReporter™ lacZ Flow Cytometry Kit

Catalog No. 501121520
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121520 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121520 Supplier Invitrogen™ Supplier No. F1930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FluoReporter™ lacZ Flow Cytometry Kit

The FluoReporter™ lacZ Flow Cytometry Kit provides materials and protocols for quantitating β-galactosidase activity with fluorescein di-V-galactoside (FDG) in single cells using flow cytometry. This kit is accompanied by a license to practice patented techniques for loading FDG by hypotonic shock and improving retention of fluorescein in lacZ-positive cells.

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Fluorescence
Content And Storage Contains 1 vial FDG (0.5 mL, 20 mM in water/DMSO/ethanol, 8:1:1, v/v), 1 vial PETG (2.0 mL, 50 mM in water), chloroquine diphosphates (1.0 mL, 30 mM in water) and 1 vial of propidium iodide (1.0 mL, 150 μM in water).

Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line FluoReporter
Quantity 1 Kit
Form Solution

For Research Use Only. Not for use in diagnostic procedures.