missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-9-acetic acid, 99+%

Catalog No. AAA1877203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1877203 1 g
AAA1877206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1877203 Supplier Thermo Scientific Chemicals Supplier No. A1877203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6284-80-6 | C15H12O2 | 224.259 g/mol

It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 6284-80-6
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00013262
InChI Key WFSMJMTYIMFHPV-UHFFFAOYSA-N
Synonym 9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid
PubChem CID 94852
IUPAC Name 2-(9H-fluoren-9-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 224.26
Percent Purity ≥99.5%
Chemical Name or Material Fluorene-9-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only