accessibility menu, dialog, popup

UserName

CAS RN 6284-80-6

IUPAC Name: 2-(9H-fluoren-9-yl)acetic acid
Synonyms: 9H-fluoren-9-ylacetic acid
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: WFSMJMTYIMFHPV-UHFFFAOYSA-N
SMILES: OC(=O)CC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)