missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoranthene 98.0+%, TCI America™
$68.97 - $713.94
Chemical Identifiers
| CAS | 206-44-0 |
|---|---|
| Molecular Formula | C16H10 |
| Molecular Weight (g/mol) | 202.26 |
| MDL Number | MFCD00001184 |
| InChI Key | GVEPBJHOBDJJJI-UHFFFAOYSA-N |
| Synonym | idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene |
| PubChem CID | 9154 |
| ChEBI | CHEBI:33083 |
| IUPAC Name | fluoranthene |
| SMILES | C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Chemical Identifiers
| 206-44-0 | |
| 202.26 | |
| GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
| 9154 | |
| fluoranthene |
| C16H10 | |
| MFCD00001184 | |
| idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
| CHEBI:33083 | |
| C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Specifications
| 206-44-0 | |
| Yellow | |
| C16H10 | |
| 25 g | |
| GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
| fluoranthene | |
| 9154 | |
| 202.26 | |
| Crystalline Powder |
| 110°C | |
| 384°C | |
| MFCD00001184 | |
| idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
| C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 | |
| 202.26 | |
| CHEBI:33083 | |
| ≥98.0% (GC) | |
| Fluoranthene |
Safety and Handling
EINECSNumber : (4)-0002
RTECSNumber : LL4025000
TSCA : Yes