CAS RN 206-44-0
IUPAC Name:
fluoranthene
Synonyms:
fluoranthene 1,2-benzacenaphthene 1,2-(1,8-naphthylene)benzene benzo(jk)fluorene idryl rcra waste number U120 1,2-(1,8-naphthylene)-benzene
Molecular Weight (g/mol):
202.26
Molecular Formula:
C16H10
InChi Key:
GVEPBJHOBDJJJI-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13
1
–
1
of
1
results
1
Fluoranthene 98.0+%, TCI America™
CAS: 206-44-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.26 MDL Number: MFCD00001184 InChI Key: GVEPBJHOBDJJJI-UHFFFAOYSA-N Synonym: idryl,1,2-benzacenaphthene,benzo jk fluorene,rcra waste number u120,1,2-1,8-naphthylene benzene,benzene, 1,2-1,8-naphthalenediyl,1,2-1,8-naphthalenediyl benzene,benzene, 1,2-1,8-naphthylene,benzo j,k fluorene PubChem CID: 9154 ChEBI: CHEBI:33083 IUPAC Name: fluoranthene SMILES: C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13