missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoranthene 98.0+%, TCI America™
Supplier: TCI America F0016100G
Specifications
| Fluoranthene | |
| 110°C | |
| 384°C | |
| MFCD00001184 | |
| idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
| C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 | |
| 202.26 | |
| CHEBI:33083 | |
| ≥98.0% (GC) |
| 206-44-0 | |
| Yellow | |
| C16H10 | |
| 100 g | |
| GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
| fluoranthene | |
| 9154 | |
| 202.26 | |
| Crystalline Powder |
Chemical Identifiers
| 206-44-0 | |
| 202.26 | |
| GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
| 9154 | |
| fluoranthene |
| C16H10 | |
| MFCD00001184 | |
| idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
| CHEBI:33083 | |
| C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Safety and Handling
EINECSNumber : (4)-0002
RTECSNumber : LL4025000
TSCA : Yes