missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Flunixin meglumine
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Solubility | Solubility aq. soln.: soluble. Other solubilities: soluble in dmso and methanol |
| Melting Point | 135.0°C to 137.0°C |
| Formula Weight | 491.46 |
| InChI Key | MGCCHNLNRBULBU-WZTVWXICSA-N |
| SMILES | CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC1=C(NC2=NC=CC=C2C(O)=O)C=CC=C1C(F)(F)F |
| IUPAC Name | (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; 2-{[2-methyl-3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 491.46 |
| Chemical Name or Material | Flunixin meglumine |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 255-836-0 |
RUO – Research Use Only