Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
$97.69 - $186.77
Specifications
Product Type Pump Set
Tubing Size 10 in. TPE 374, 19.7 in. Pharmed BPT
View More Specs
Products 5
Catalog Number Mfr. No. Tubing Size Connector Types Dimensions Price Quantity  
Catalog Number Mfr. No. Tubing Size Connector Types Dimensions Price Quantity  
SH3111101
View Documents
Thermo Scientific™
SH3111101
10 in. TPE 374, 19.7 in. Pharmed BPT Luer 1/8 in. ID x 1/4 in. OD
Each for $97.69
Only null left
SH3111201
View Documents
Thermo Scientific™
SH3111201
10 in. TPE 374, 19.7 in. Pharmed BPT MPC 3/8 in. ID x 5/8 in. OD
Each for $115.64
Only null left
SH3111301
View Documents
Thermo Scientific™
SH3111301
10 in. TPE 374, 19.7 in. Pharmed BPT MPX 1/2 in. ID x 3/4 in. OD
Each for $186.77
Only null left
SH3111501
View Documents
Thermo Scientific™
SH3111501
10 in. TPE 374, 19.7 in. Pharmed BPT MPC 1/4 in. ID x 7/16 in. OD
Each for $106.53
Only null left
SH3111401
View Documents
Thermo Scientific™
SH3111401
10 in. TPE 374, 19.7 in. Pharmed BPT MPC 1/4 in. ID x 3/8 in. OD
Each for $101.91
Only null left
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
Pump Set
10 in. TPE 374, 19.7 in. Pharmed BPT
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'