Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Supplier:  Thermo Scientific™ SH3111301
 View more versions of this product
Catalog No. SH3111301
Only null left
Explore available promotions
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
1/2 in. ID x 3/4 in. OD
MPX
10 in. TPE 374, 19.7 in. Pharmed BPT
12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m (10 in.); TPE 374; MPX
Pump Set
Product Suggestions
Product Suggestions