Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Supplier: Thermo Scientific™ SH3111101
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
| 1/8 in. ID x 1/4 in. OD | |
| Luer | |
| 10 in. TPE 374, 19.7 in. Pharmed BPT |
| 3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m(10 in.); TPE 374; Luer | |
| Pump Set |