missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flufenamic acid, 97%

Catalog No. AC165920500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165920500 50 g Glass bottle
AC165920100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC165920500 Supplier Thermo Scientific Chemicals Supplier No. 165920500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 530-78-9 | C14H10F3NO2 | 281.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 530-78-9
Molecular Formula C14H10F3NO2
Molecular Weight (g/mol) 281.22
MDL Number MFCD00002422
InChI Key LPEPZBJOKDYZAD-UHFFFAOYSA-N
Synonym flufenamic acid, fluphenamic acid, nichisedan, achless, arlef, flufacid, fullsafe, lanceat, paraflu, plostene
PubChem CID 3371
ChEBI CHEBI:42638
IUPAC Name 2-[3-(trifluoromethyl)anilino]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F

Specifications

Melting Point 132°C to 135°C
Color Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula 2-(CF3C6H4NH)C6H4CO2H
Quantity 50 g
Beilstein 14, III, 905
Merck Index 15, 4163
Solubility Information Solubility in water: 0.0265g/L (37°C). Other solubilities: soluble in ethanol (6.9g/L at 37°C),chloroform,(19g/L at 37°C),dmso,cyclohexane
Formula Weight 281.22
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Flufenamic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroug
EINECSNumber 208-494-1
RTECSNumber CB4375000

RUO – Research Use Only