missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fludarabine Monophosphate 98.0+%, TCI America™
Supplier: TCI America F0913100MG
Specifications
| Fludarabine Monophosphate | |
| 203°C | |
| C10H13FN5O7P | |
| 100 mg | |
| GIUYCYHIANZCFB-FJFJXFQQSA-N | |
| {[(2R,3S,4S,5R)-5-(6-amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid | |
| 30751 | |
| 365.21 | |
| Crystalline Powder |
| 75607-67-9 | |
| White-Yellow | |
| MFCD00866418 | |
| fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine monophosphate, famp, unii-1x9vk9o1sc | |
| NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC(F)=N1 | |
| 365.21 | |
| CHEBI:63599 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 75607-67-9 | |
| 365.21 | |
| GIUYCYHIANZCFB-FJFJXFQQSA-N | |
| 30751 | |
| {[(2R,3S,4S,5R)-5-(6-amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid |
| C10H13FN5O7P | |
| MFCD00866418 | |
| fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine monophosphate, famp, unii-1x9vk9o1sc | |
| CHEBI:63599 | |
| NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC(F)=N1 |
Safety and Handling
RTECSNumber : UO7440900
TSCA : No