missing translation for 'onlineSavingsMsg'
Learn More

Fludarabine Monophosphate 98.0+%, TCI America™

Catalog No. p-7129974
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F091325MG 25 mg
F0913100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F091325MG Supplier TCI America Supplier No. F091325MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75607-67-9
Molecular Formula C10H13FN5O7P
Molecular Weight (g/mol) 365.21
MDL Number MFCD00866418
InChI Key GIUYCYHIANZCFB-FJFJXFQQSA-N
Synonym fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine monophosphate, famp, unii-1x9vk9o1sc
PubChem CID 30751
ChEBI CHEBI:63599
IUPAC Name {[(2R,3S,4S,5R)-5-(6-amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC(F)=N1

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 25 mg
Formula Weight 365.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fludarabine Monophosphate
RTECSNumber UO7440900
TSCA No