missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Fluconazole | |
| 141°C | |
| C13H12F2N6O | |
| fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol | |
| C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O | |
| 306.277 | |
| CHEBI:46081 | |
| ≥98.0% (GC,T) |
| 86386-73-4 | |
| White-Yellow | |
| 1 g | |
| RFHAOTPXVQNOHP-UHFFFAOYSA-N | |
| 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol | |
| 3365 | |
| 306.28 | |
| Crystalline Powder |
Chemical Identifiers
| 86386-73-4 | |
| 306.277 | |
| fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol | |
| CHEBI:46081 | |
| C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O |
| C13H12F2N6O | |
| RFHAOTPXVQNOHP-UHFFFAOYSA-N | |
| 3365 | |
| 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol |
Safety and Handling
RTECSNumber : XZ4810000
TSCA : No