accessibility menu, dialog, popup

UserName

CAS RN 86386-73-4

IUPAC Name: 2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Synonyms: fluconazole fluconazole product fluconazol abz fluzone flucazol fluconazolum biocanol lichtenstein brand of fluconazole apo-fluconazole fluc hexal
Molecular Weight (g/mol): 306.28
Molecular Formula: C13H12F2N6O
InChi Key: RFHAOTPXVQNOHP-UHFFFAOYSA-N
SMILES: OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)