missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole, 98%

Catalog No. p-7077141
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC101910050 5 g
AC101910250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC101910050 Supplier Thermo Scientific Chemicals Supplier No. 101910050
Only null left

CAS: 31430-15-6 | C16H12FN3O3 | 313.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31430-15-6
Molecular Formula C16H12FN3O3
Molecular Weight (g/mol) 313.28
InChI Key CPEUVMUXAHMANV-UHFFFAOYSA-N
Synonym flubendazole, flumoxane, flubendazol, fluvermal, flubenol, flubendazolum, flumoxal, flutelmium, methyl 5-4-fluorobenzoyl-1h-benzo d imidazol-2-yl carbamate, flubendazol inn-spanish
PubChem CID 35802
ChEBI CHEBI:77095
IUPAC Name methyl N-[6-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=C(C=C3)F

Specifications

Melting Point 260.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 4 hr)
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 313.28
Chemical Name or Material Flubendazole
EINECSNumber 250-624-4

RUO – Research Use Only