accessibility menu, dialog, popup

UserName

H3COOHNHNNOF

CAS RN 31430-15-6

IUPAC Name: methyl N-[6-(4-fluorobenzoyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: flubendazole
Molecular Weight (g/mol): 313.29
Molecular Formula: C16H12FN3O3
InChi Key: CPEUVMUXAHMANV-UHFFFAOYSA-N
SMILES: COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)