missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole 98.0+%, TCI America™

Catalog No. p-7129921
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08255G 5 g
F082525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08255G Supplier TCI America Supplier No. F08255G
Only null left

Chemical Identifiers

CAS 31430-15-6
Molecular Formula C16H12FN3O3
Molecular Weight (g/mol) 313.288
MDL Number MFCD00871999
InChI Key CPEUVMUXAHMANV-UHFFFAOYSA-N
Synonym flubendazole, flumoxane, flubendazol, fluvermal, flubenol, flubendazolum, flumoxal, flutelmium, methyl 5-4-fluorobenzoyl-1h-benzo d imidazol-2-yl carbamate, flubendazol inn-spanish
PubChem CID 35802
ChEBI CHEBI:77095
IUPAC Name methyl N-[6-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=C(C=C3)F

Specifications

Melting Point 260°C
Color Yellow
Quantity 5 g
Formula Weight 313.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Flubendazole
RTECSNumber DD6497500
TSCA No