missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Floating Rack, Mega
Description
- Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
- Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
- One-step procedure that does not require syringes or any special equipment
- Sample volume recovery > 97%
- Protease, RNAse, DNAse, and PCR product free
Specifications
Specifications
| Product Type | Floating Rack, Mega |
| Quantity | 10 rack |
| Content And Storage | +15°C to +30°C |