missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Floating Centrifuge Tube Rack

For use on bench and in ice and water baths, floating microtube rack perfect for controlling sample temperature

$48.84

Specifications

Autoclavable Autoclavable
Color White
For Use With (Equipment) 1.5 or 2.0 Microtubes
Type Rack
Material Polypropylene
View More Specs

Products 2
Catalog Number Mfr. No. Shape Price Quantity & Availability  
Catalog Number Mfr. No. Shape Price Quantity & Availability  
14127131
View Documents Promotion Details
Fisherbrand™
HS2132AF2
Square
Pack of 4 Each for $48.84
Case of 4 Pack for $195.36
Only null left
 
14127184
View Documents Promotion Details
Fisherbrand™
HS2134AF2
Round
Pack of 4 Each for $48.84
Case of 4 Pack for $195.36
Only null left
 
Description

Description

  • The Floating Microtube Rack can be used on the bench, with water baths or ice containers. The rack keeps microtubes afloat in a water bath or can be pushed into crushed ice. It is perfect for controlling sample temperature
  • Made of polypropylene
  • Attach legs and rack stands upright for use on bench top
  • Detach legs and samples can be quickly removed by pushing down onto any surface
Specifications

Specifications

Autoclavable
1.5 or 2.0 Microtubes
Polypropylene
10.9 mm
White
Rack
4
Videos
SDS
Documents

Documents