missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Floating Centrifuge Tube Rack

Catalog No. 14127131
Change view
Click to view available options
Shape:
Round
Square
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
14127131 Square
14127184 Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14127131 Supplier Fisherbrand™ Supplier No. HS2132AF2

For use on bench and in ice and water baths, floating microtube rack perfect for controlling sample temperature

  • The Floating Microtube Rack can be used on the bench, with water baths or ice containers. The rack keeps microtubes afloat in a water bath or can be pushed into crushed ice. It is perfect for controlling sample temperature
  • Made of polypropylene
  • Attach legs and rack stands upright for use on bench top
  • Detach legs and samples can be quickly removed by pushing down onto any surface

Specifications

Autoclavable Autoclavable
Color White
For Use With (Equipment) 1.5 or 2.0 Microtubes
Type Rack
Holds 16 Microtubes
Material Polypropylene
No. of Positions 16
No. per Pack 4
Shape Square
Diameter (Metric) Well 10.9 mm